Cleanroom Mini-Vac Systems, Tiger-Vac®
Supplier: Tiger-Vac International
110737A 110739A 110735A
14900-022EA 4271.75 USD
14900-022 14900-024 14900-020
Cleanroom Mini-Vac Systems, Tiger-Vac®
Vacuum Cleaners
Ideal for cleanroom and laboratory maintenance, these stainless steel vacuum systems feature an ULPA (Ultra Low Penetration Air) filter with 99.999% efficiency on particles as small as 0.12µm
Three vacuum models are available: back pack, golf cart, and handheld. All units feature a compact design for easy handling and storage. The back pack unit features an ergonomic harness for maximum maneuverability and comfort. The golf cart unit features a convenient stainless steel platform for easy mobility. The handheld unit can be carried or placed on or under most tables; an optional shoulderMore Product Information