VWR® Support Jacks, Stainless Steel
Supplier: VWR International
80863301 80863304 80863310 80863313 80863316 80863319 80863307
14233-360EA 239.19 USD
14233-360 14233-362 14233-366 14233-368 14233-370 14233-372 14233-364
VWR® Support Jacks, Stainless Steel
Laboratory Jacks
Stainless steel lab jacks are ideal for use in fume hoods or benchtops.
  • Stainless steel construction
  • Autoclavable and chemical-resistant
  • Seven convenient sizes
These ultra-stable lifting platforms hold a variety of items such as glassware, hot plates, baths, and magnetic stirrers
Ratchet tool is designed to add extra leverage to the 30.5x30.5 cm (12x12") and 40.6x40.6 cm (16x16") support jacks. The ratcheting handle easily attaches to the actuating rod to allow easy, accurate adjustments.
Support rod kit includes a 43.2 cm (17") vertical supportMore Product Information