Samco™ Bio-Tite™ Specimen Container for Operating Room, Thermo Scientific
Supplier: Samco
07 0006
14231-916CS 315.36 USD
14231-916
Samco™ Bio-Tite™ Specimen Container for Operating Room, Thermo Scientific
Sample Containers General Purpose Containers
Samco™ Bio-Tite™ Specimen containers for the operating room.
  • Supplied sterile
  • Shatter-resistant containers are manufactured from virgin polypropylene
  • Leak-resistant seal prevents bio-hazardous spills
Container is ideal for collecting biopsy specimens and transporting them into a sterile environment. It is tested at varying pressures, making it ideal for use in various transport systems.
Certificaciones: FDA Class 1 Exempt Medical Device.
Información para pedidos: Ambient storage and shipping recommended for containers.