Compact Roller System for Small Bottles, Wheaton®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W348930-CH W348924-A
14231-390CS 632.57 USD
14231-390 14231-392
Compact Roller System for Small Bottles, Wheaton®, DWK Life Sciences
Roller Culture Apparatus
Compact unit is ideal for benchtop use
Designed to roll vessels that are 108–121mm in diameter and up to 290mm long. Two outside rollers move inward to fit vessels with diameters as small as 75mm. The single-deck unit accommodates two bottles. Use in CO2 environments may necessitate more frequent replacement of motor brushes. Up to two additional deck kits can be added to accommodate an additional two bottles per kit.
Certificaciones: UL listed. CSA certified. CE marked.
Información accesorios: Additional deck kit for small bottle bench-top roller rack allows for double or tripleMore Product Information