Potter-Elvehjem Tissue Grinders, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
357984 357989 357994 357979 358028 358033 357969 357974 358043 358053 358029 358034 358038 358039 358036 358044 358049 358054 357844
14231-368CS 439.74 USD
14231-368 14231-370 14231-372 14231-384 14231-340 14231-342 14231-364 14231-366 14231-374 14231-376 62400-700 62400-722 62400-733 62400-744 62400-755 62400-766 62400-777 62400-788 62400-799 62400-802 62400-879
Potter-Elvehjem Tissue Grinders, Wheaton
Homogenizers Tissue Grinders
Grinders for preparation of soft tissue homogenates, such as liver, brain.
  • Grinders are available with smooth pestles or serrated pestles, which disperse homogenate into the mortar cylinder more effectively
  • Depth of radial serrations is 0.508–0.635 mm (0.020–0.025")
  • Smooth or serrated pestles are available
  • Autoclavable and parts are interchangeable
All pestles are made of fluoropolymer resin with a 6.4 mm (1/4") diameter stainless steel shaft. Radius at pestle bottom conforms to radius in bottom of grinding chamber. Grinding chamber is made of lowMore Product Information