Histosette® II Tissue Processing/Embedding Cassettes, Simport
Supplier: Simport Scientific
M485-2 M485-12 M485-6 M485-10 M485-3 M485-4 M485-8 M485-9 M485-5 M485-7 M485-11
14230-470CS 296.98 USD
14230-470 14230-490 14230-478 14230-486 14230-472 14230-474 14230-482 14230-484 14230-476 14230-480 14230-488
Histosette® II Tissue Processing/Embedding Cassettes, Simport
Embedding Cassettes Tissue Processing Cassettes
These disposable cassettes hold tissue specimens efficiently during the embedding process and in storage
Molded from acetal polymer, they keep specimens submerged in liquid and are resistant to the chemical action of histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. Anterior writing area is angled at 45° for suitability with certain types of cassette labeling instruments. Colors are non-cytotoxic and nonmetallic
Información para pedidos: Lids are supplied unattached. Packed in two dispenser boxes of 500 cassettes and oneMore Product Information