MµltiFit™ Universal Fit Pipette Tips, Sorenson BioScience
Supplier: Sorenson Bioscience
10190T 10200T 10230T 10470T 15270T 10520T 15370 15660T 15670 15670T 15720 15720T 15730 30430T 30440 30450T 10620 10660 10590 10500 10520 10470 10200 10230 10080 10190 10130 10040
14229-740CS 535.2 USD
14229-740 14229-742 14229-744 14229-746 14229-748 14229-750 14229-796 14229-822 14229-824 14229-826 14229-828 14229-830 14229-832 14229-866 14229-868 14229-872 14229-874 14229-876 14229-886 14229-888 14229-894 53550-065 53550-076 53550-087 53550-090 53550-093 53550-094 53550-096 53550-099 53550-101 53550-103 53550-113 53550-123
MµltiFit™ Universal Fit Pipette Tips, Sorenson BioScience
Pipette Tips
Pipette tips feature a unique beveled end to minimize fluid retention.
  • Fully autoclavable
  • Tips fit most popular research-grade pipettors
They ensure superior sample delivery and recovery in critical micro-volume ranges. Reinforced base design and standard 96-well pattern of cell racks ensures rapid mounting of small tips on all multichannel pipettors.
Certificaciones: The 1 to 200 µl racked tips are certified RNase-, DNase-, human DNA-, PCR inhibitor-, and pyrogen-free.
Información para pedidos: Available in bulk packaging, autoclavableMore Product Information