Unions for Gas Chromatography, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
101230 101270 101290 101300 101340 103330 103400 103403 1034030 103404 103405 103406 103407 103408 103418 1034180 1034183 1034187 1034604 1034605 103461 103420 1034200 18500002 103402 1034050 1236300 1236302 1236306
14226-500EA 42.22 USD
14226-500 14226-504 14226-506 14226-508 14226-514 14226-518 14226-520 14226-522 14226-524 14226-526 14226-528 14226-530 14226-532 14226-534 14226-538 14226-540 14226-544 14226-546 14226-566 14226-568 14226-570 14227-500 14227-502 10799-798 10799-800 10799-802 10799-804 10799-806 10799-808
Unions for Gas Chromatography, Trajan Scientific and Medical
Fittings and Connectors
Stainless steel unions, tees and crosses come in a variety of tube sizes and configurations, such as straight through, reducing, tees, crosses and bulkheads
GLT unions are inert and mainly used for connecting fused silica tubing of various internal diameters. All SGE unions are supplied with the appropriate nuts, ferrules and/or sealing rings. Replacement nuts are also available.
Certificaciones: RoHS compliant per Directive 2011/65/EU
Información para pedidos: Unions and nuts are available in a range of sizes to connect 1/4, 1/8, and 1/16" tubing and fused silica.
Order Now
SPECIFICATIONS
Environmentally Preferable