Trajan, SilTite™ Mini-Unions, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
073470 073471 073472 073550 073551 073552 073553
14226-430PK 166.13 USD
14226-430 14226-432 14226-434 14226-436 14226-438 14226-440 14226-442
Trajan, SilTite™ Mini-Unions, Trajan Scientific and Medical
Fittings and Connectors
Your chromatography analysis does not end with the selection of the GC column.
  • Inert and precise glass lined bore
  • Robust and stable
  • For both extreme temperature and pressure cycling
  • Leak-free
  • No need to re-tighten after installation
  • Low dead volume
These unions should be used when fitting a pre-column or guard column, GC to GC connections. They can be used to repair broken columns.
The combination of components selected for your instrument also make an important contribution to successful separations. SGE brings technology and chromatographyMore Product Information