TWD Scientific Weighing Funnels
Supplier: TWD Scientific
GWF-BS-XS DPWF-PP-XS ASWF4XL ASWF3L ASWF2M ASWF1S ASWFXS DPWF-PP4-XL DPWF-PP3-L DPWF-PP2-M DPWF-PP1-S GWF-BS4-XL GWF-BS3-L GWF-BS2-M GWF-BS1-S
14221-800CS 1627.78 USD
14221-800 14221-802 14221-804 14221-806 14221-808 14221-810 14221-812 14221-814 14221-816 14221-818 14221-820 14221-822 14221-824 14221-826 14221-828
TWD Scientific Weighing Funnels
Boats Weighing Boats
The TWD Scientific (formerly Tradewinds Direct) family of weighing funnels (weighing boats) all feature a flat bottom for sitting on balances and a pouring neck that fits to volumetric flasks, micro-tubes, or any narrow neck apparatus for spill-free transfer.
  • Available in antistatic polypropylene, disposable polypropylene, and borosilicate glass
  • Residual samples can be rinsed to guarantee 100% transfer
  • Five different sizes of funnels fit to various flask sizes
  • X-Small funnels fit 96-well mini-tubes and deep well plates
Suitable for safe handling of hazardous materials.