Eyesaver® Deck-Mount Swing-Out Eyewash, Speakman®
Supplier: Speakman
SE-570
14214-370EA 1265.93 USD
14214-370
Eyesaver® Deck-Mount Swing-Out Eyewash, Speakman®
First Aid Supplies Eyewash Stations and Eyewash Bottles
Low-profile counter-top eyewash unit for installations where space is at a premium.
  • Unit is activated when rotated into place
Features twin aerated sprays, stay-open ¹/₄-turn Perfect Valve* with swing activation, polished chrome finish, left or right hand operation for installation flexibility, built-in flow regulator to compensate for varying supply pressures, and deck/wall mounted sign for easy identification.
Certificaciones: Meets ANSI Z358.1 standard.