SecureSeal™ Adhesive Film for Microplates, Simport Scientific
Supplier: Simport Scientific
T329-3
14211-710CS 1324.09 USD
14211-710
SecureSeal™ Adhesive Film for Microplates, Simport Scientific
Seals Microplate Seals
This adhesive sealing films reduce sample-to-sample or well-to-well contamination and or spill over.
  • Compatible with almost all microplate procedures
  • RNase, DNase, DNA-free
  • DMSO resistant
  • Temperature range: –40...+120 °C
The polyester based film with acrylic adhesive is inert and thus compatible with almost all microplate procedures. SecureSeal™ is less than .001% moisture permeable under high humidity conditions and less than .01% oxygen permeable.
Envase: Each package contains 100 sheets.