SP Wilmad-LabGlass Straight Powder Funnels, SP Industries
Supplier: SP Industries
LG-8300-110
14202-028EA 92.86 USD
14202-028 14202-386 14202-768 14206-388 14208-348
SP Wilmad-LabGlass Straight Powder Funnels, SP Industries
Funnels Powder Funnels
These straight funnels are used for the introduction of liquids or powders into containers with the same Standard Taper joint.
The funnel design promotes free flow and provides a smooth surface for the transfer of material.
Order Now
SPECIFICATIONS
Environmentally Preferable