SP Wilmad-LabGlass Barrett Moisture Traps, SP Industries
Supplier: SP Industries
LG-9075-102
14201-042EA 653.11 USD
14201-042 14212-730
SP Wilmad-LabGlass Barrett Moisture Traps, SP Industries
Distillation Components Distillation Receivers
Calibrated “to contain” at 20°C, these distilling receivers are used for determination of water in various materials by distillation with a solvent that is not miscible with water.
  • Borosilicate glass
With full length Standard Taper inner member for connection to distilling flask and a Standard Taper outer joint of the same size to connect the condenser, these moisture traps are supplied with a 2mm stopcock.