PolarPack® Gel Packs, Ice Pack Refrigerant Gel Packs, Sonoco ThermoSafe
Supplier: Sonoco Thermosafe
PP16 PP24 PP3 PP48 PP12 PP32 PP6 PP8 TUFF24 PP20IN24BL PP42 PP8IN24BAGS
14100-476CS 27.67 USD
14100-476 14100-478 16000-068 82029-734 89024-674 89049-998 89050-000 89050-002 89495-996 89495-998 10770-550 75845-138
PolarPack® Gel Packs, Ice Pack Refrigerant Gel Packs, Sonoco ThermoSafe
Cooling Packs
An economical alternative to wet ice packs, these nontoxic, reusable gel packs are enclosed in a durable 4.1 mil polyethylene film.
  • Reliable non-mechanical refrigeration
  • Ideal for transporting food and pharmaceuticals
  • Economical alternative to wet ice
PolarPack gel packs provide reliable non-mechanical refrigeration for the safe transport of food products, pharmaceuticals, and other medical products
The elongated design of PP8IN24BAGS allows for a protective wrap around sensitive specimen samples and diagnostic materials in vials and tubes duringMore Product Information