DA-7645® Process and Research Cleaners
Supplier: Steris
116505 116501 1165D3
14004-166EA 587.23 USD
14004-166 14004-168 77531-102
DA-7645® Process and Research Cleaners
Cleaning Agents Detergents and Surface Cleansers
DA-7645® Detergent is a proprietary blend of surfactants, solvents and other performance-enhancing ingredients that provide multiple cleaning mechanisms in manual cleaning applications.
  • Efficacy - Mild pH formulation delivers excellent cleaning characteristics and substrate compatibility with enhanced operator safety
  • Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports
  • Productivity - Delivers efficiency to
More Product Information