Cage-Klenz® 280 Acid-Based Detergents
Supplier: Steris
1K2802 1K2805
14004-122EA 1541.7 USD
14004-122 77531-220
Cage-Klenz® 280 Acid-Based Detergents
Cleaning Agents Detergents and Surface Cleansers
Cage-Klenz® 280 Detergent is a phosphoric acid-based detergent for use in high pressure spray animal cage washing equipment.
  • Efficacy - Effectively cleans hard water and urine scale, and metal oxides in both soft and hard water
  • Flexibility - Excellent materials compatibility with a wide variety of caging materials including polycarbonate
  • Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Environmentally Friendly - formulation with biodegradable surfactants
  • Reliability - Manufactured in a cGMP
More Product Information