Nalgene® Graduated Transparent Beakers with Handle, PMP, Thermo Scientific
Supplier: Thermo Fisher Scientific
1223-1000 1223-3000
13916-988CS 492.92 USD
13916-988 13916-999
Nalgene® Graduated Transparent Beakers with Handle, PMP, Thermo Scientific
Beakers
Polymethylpentene (PMP) material allows a clear view of beaker contents.
  • Rectangular handle for easy carrying and pouring
  • Molded-in graduations in both mL and oz. will not wear off with continuous use or washings
Excellent chemical resistance to acids, bases and aliphatic alcohols and useful for a broad range of solutions. Withstand continuous exposure to temperatures as high as 150°C (302°F) and intermittent temperatures to 175°C (347°F).
Order Now
SPECIFICATIONS
Environmentally Preferable