ErgoSCRUB® Scrubbing Handles and ScrubDISK® Pads, Foamtec International
Supplier: Foamtec International
FT901 FT951 FT950 HT4518DC3-1 HT4528DC3-1 HT4536DC3G-1 HT4518DC3G-1 HT4536DC3-1 HT4522DC3-1 HT4580DC3-1 FT951 HT4536DC3G-1
13501-880CS 261.27 USD
13501-880 13501-882 16003-844 10811-792 10811-794 10811-802 16003-838 89069-036 89009-519 16003-475 89133-224 10811-803
ErgoSCRUB® Scrubbing Handles and ScrubDISK® Pads, Foamtec International
Wipes Cleaning Kits and Pads
Ergonomically correct, disposable, and reusable scrubbing handles and pads for cleaning critical surfaces on process equipment in the semiconductor, flat-panel display, optical component, and hard disk drive industries.
  • Pads contain diamond or silicon carbide grit
  • Hard handle scrubber is for use on flat surfaces such as Novellus™ heat blocks
Silicon carbide pads are backed with a pressure-sensitive adhesive for use with the disposable handles. Diamond pads are backed with either a pressure-sensitive adhesive for attachment to the disposable handles orMore Product Information