SP Bel-Art Curved Forceps, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H37940-0000
13197-442BG 85.99 USD
13197-442
SP Bel-Art Curved Forceps, Bel-Art Products, a part of SP
Forceps
The exceptionally fine tips of these forceps are coated with brown Fluo-Kem® Teflon® FEP which complies with FDA regulations covering direct contact with food.
  • FEP coating
  • Serrated, curved tips
  • Suited for grasping fine objects
They have an overall length of 9.8 cm (3⅞") and serrated, curved tips.