VWR® Advanced and Standard Multi-Position Stirrers, 230 V (Export Only)
Supplier: VWR International
12621-020 12621-028 12621-036 12621-044 12621-048
12621-020EA 2003.78 USD
12621-020 12621-028 12621-036 12621-044 12621-048
VWR® Advanced and Standard Multi-Position Stirrers, 230 V (Export Only)
Stirrers Magnetic Stirrers, Multi-position
Microprocessor controlled
Synchronized and uniform mixing
Low-profile design
Best-in-Class, 5-year warranty
Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner.
  • 230 V model
  • Precise speed control
  • Cast metal base offers durability and added stability
  • Excellent low-speed stirring
Standard series offers an analog speed control knob with dial markings
Powerful magnetic stirrers with precise speed control offer repeatable performance. Synchronized operation ensures uniform mixing at each position. Ramping featureMore Product Information