VWR® Dylatherm™ Hotplate 230 V
Supplier: VWR International
12620-980
12620-980EA 369.35 USD
12620-980
VWR® Dylatherm™ Hotplate 230 V
Heaters Hot Plates
The economical Dylatherm™ Hot Plate features a large 16.5 cm (6.5") diameter cast aluminum top plate that provides an even heating surface capable of heating 1500 mL of water.
  • Large 16.5 cm (6.5") diameter cast aluminum top plate
  • Built-in support rod holder
Bimetallic thermostat offers reliable and repeatable temperature control
Built-in support rod holder with locking knob accepts optional support rod and clamp kit for mounting thermometers and temperature probes. Kit includes 45.7 cm (18”) support rod, thermometer extension clamp, three-prongMore Product Information