VWR® Signature™ Incubating 3-D Rotator Waver
Supplier: VWR International
12620-920 10027-114
12620-920EA 4737.92 USD
12620-920 10027-114
VWR® Signature™ Incubating 3-D Rotator Waver
Shakers and Mixers Shaking Incubators
Incubating 3-D Rotator Waver combines the unique vertical and horizontal wave motion with general-purpose incubation into one compact unit.
  • Vertical and Horizontal Orbital Motion
  • Digital Display for Speed, Temperature, Tilt Angle, and Time
  • Microprocessor Control
  • Timer with Audible Alarm
  • Compact, Benchtop Design
  • Now with Best-in-Class, Exclusive 5-Year Warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
Microprocessor PID temperature controller maintains precise control and can be set in 1°More Product Information