Whirl-Pak® Bags for Seward Stomacher® Lab Blenders, Nasco®
Supplier: NASCO SAMPLING LLC
B01195 B01196
11216-777CS 708.19 USD
11216-777 11216-280
Whirl-Pak® Bags for Seward Stomacher® Lab Blenders, Nasco®
Bags Blender Bags
Sterile Whirl-Pak™ bags are designed to keep liquid, semi-solid, or solid samples in the bag during blending.
  • All bags have a continuous perimeter seal
Bags are made of heavy-duty 4 mil-thick polyethylene, except for 11216-796, which is made of heavy-duty 3 mil-thick polyethylene. Bags feature patented Puncture Proof Tabs with tape extending past the wire ends to eliminate sharp points that could puncture the bag or cause damage to skin or gloves.
Información para pedidos: Selected styles incorporate a white, write-on strip for labeling. SizesMore Product Information