Double-Sided Cleanroom Tape, Polyester, Ultratape
Supplier: Ultratape Industries
1510CL050P3D 1510CL100P3D 1510CL200P3D 1520CL050P3D 1520CL100P3D 1520CL200P3D
11212-400CS 2266.71 USD
11212-400 11212-404 11212-406 11212-420 11212-424 11212-426
Double-Sided Cleanroom Tape, Polyester, Ultratape
Tapes
This cleanroom tape is ideal for signs, nameplate mounting, or for connecting plastic sheeting pieces in cleanroom wall construction.
Choose removable or permanent adhesive double-sided tape. Removable adhesive tapes have no liner, for use in a cleanroom tape dispenser. Will not leave residue. Permanent adhesive tapes are clear with a disposable red Mylar® liner. Will leave residue.
For dispenser, see 11212-142 series.
Order Now
SPECIFICATIONS
Environmentally Preferable