Miniature Air Sampling Kits (Stainless Steel and Siltek®-Treated), Restek
Supplier: Restek
26258 26252 26256 26253 26257 26259 26254
10862-612EA 1258.21 USD
10862-612 10862-872 10862-874 10862-970 10862-974 10862-976 10863-030
Miniature Air Sampling Kits (Stainless Steel and Siltek®-Treated), Restek
Samplers Air Samplers
Restek’s passive air sampling kit incorporates all the hardware necessary to collect air samples and is easy to assemble for field sampling.
  • Provide accurate integrated sampling without a sampling pump
  • Convenient smaller size connects easily to miniature canisters
  • Available in stainless steel or with Siltek® treatment for greater inertness
Información accesorios: Air sampling canisters sold separately.
Información para pedidos: Kit includes flow controller, critical orifice, 2 µm frit filter, vacuum gauge, and sample inlet. The gauge (cat.# 24120)More Product Information