Forced Air Circulation Incubators, Memmert
Supplier: Memmert
IF55, 230 V IF55PLUS115 IF55PLUS230 IF75, 115 V IF75, 230 V IF75PLUS115 IF75PLUS230 IF260, 115 V IF260, 230 V IF260PLUS115 IF110, 115 V IF110, 230 V IF110PLUS115 IF110PLUS230 IF160, 115 V IF160, 230 V IF160PLUS115 IF160PLUS230 IF30, 115 V IF30, 230 V IF30PLUS115 IF30PLUS230 IF55, 115 V IF750PLUS115 IF750PLUS230 IF260PLUS230 IF450PLUS115 IF450PLUS230 IF750115V IF750230V IF450115V IF450230V
10028-910EA 3741.65 USD
10028-910 10028-912 10028-914 10028-916 10028-918 10028-920 10028-922 10028-924 10028-926 10028-928 10029-040 10029-042 10029-044 10029-046 10029-048 10029-050 10029-052 10029-054 10029-096 10029-098 10029-100 10029-102 10029-104 10861-316 10861-318 10861-320 10861-454 10861-456 10861-312 10861-314 10861-322 10861-324
Forced Air Circulation Incubators, Memmert
Incubators Standard Incubators
The Generation 2012 range has natural and forced convection models with a temperature range up to +80 °C
SingleDISPLAY models; ideal for standard applications. ControlCOCKPIT parameters: Temperature (Celsius or Fahrenheit), fan speed, exhaust air flap position, programme time, time zones and daylight-saving time. One PT100 temperature sensor in a 4-wire circuit and ethernet interface for reading out the protocol log. Double over-temperature protection including visual alarm: Electronic temperature monitoring with freely adjustable monitoring temperature and mechanical temperature limiterMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Temperature Range Ambient +5…80 °C