Reversing Digital Overhead Stirrer, Caframo
Supplier: Caframo
BDC2010 BDC2010AU BDC2010U1
97001-136CS 10524.96 USD
97001-136 10860-016 10860-290
Reversing Digital Overhead Stirrer, Caframo
Stirrers Overhead Stirrers
The Reversing Digital stirrer maximizes productivity, fits into any area of the laboratory including a fume hood or enclosed workstation, and provides accurate and repeatable results every time.
This stirrer also offers a choice of rotation direction - clockwise, counter clockwise or both (agitation mode) and features a digital display, a countdown timer and a cycle timer.
Suitable for a wide range of mixing applications, this Caframo benchtop overhead stirrers are used in chemical reactions and synthesis, raw material QC, food sample prep, polymer and personal care formulations.
Order Now
SPECIFICATIONS
Environmentally Preferable