Chemical Resistant Polycarbonate Face Shields, Oberon
Supplier: Sellstrom
2128R 028 128R 128AFR
10856-378PK 531.76 USD
10856-378 10856-536 10856-538 67200-101
Chemical Resistant Polycarbonate Face Shields, Oberon
Face Shields
Lightweight and durable design protects users from potentially harmful chemicals.
  • Equipped with ratchet headgear
  • Coated by Silicote-6, to protect polycarbonate from many mild acids, bases, and solvents
Chemical resistant polycarbonate face shields are available with a variety of coatings. The replacement shield is designed for use with ratchet headgear and slot-fit adapters.