Precleaned Volatile Organic Analyte, VOA Sampling Vials, Restek
Supplier: Restek
21796 21799 26122 26121 21797 21797-220 21798
10803-256PK 164.82 USD
10803-256 10803-258 10015-256 10015-438 10854-098 10854-100 10854-102
Precleaned Volatile Organic Analyte, VOA Sampling Vials, Restek
Vials
Precleaned volatile organic analyte sampling vials are cleaned and assembled before delivery, and are lot traceable for guaranteed quality.
  • Screw-thread or vial format
  • Clear or amber color
  • Glass fabrication
  • 24 mm orifice
Volume capacity ranges from 20 to 60 ml depending on model. These vials are cleaned to U.S. EPA Protocol B specifications.
Order Now
SPECIFICATIONS
Environmentally Preferable