Plunger Seals for Shimadzu HPLC Systems, Restek
Supplier: Restek
24981 24980 25290 24985
10059-336EA 113.79 USD
10059-336 10853-432 10853-442 10853-456 10853-682
Plunger Seals for Shimadzu HPLC Systems, Restek
Seals Mechanical Seals
Graphite-filled PTFE; best for organic solvents
Ultra-high molecular weight polyethylene (UHMWPE); increased resistance to abrasion, best for buffers.
Order Now
SPECIFICATIONS
Environmentally Preferable