Anti-Mac-3 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: M3/84]

Supplier: Biolegend
108504
108504-BLEA 347.9 USD
108504-BL
Anti-Mac-3 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: M3/84]
Antibodies
FITC anti-mouse CD107b (Mac-3) [M3/84]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Mac-3
Clonality: Monoclonal
Clone: M3/84
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR