Rtx®-1 GC Columns, Restek
Supplier: Restek
10108 10121 10143 10105 10106 10107 10109 10111 10112 10120 10123 10124 10135 10138 10139 10141 10122 10125 10126 10127 10136 10140 10142 10144 10128 10137 40101 40110 40111 40112 41101 41102 41103 41104 10110 10123-124 10120-124 10124-125 10126-124 10136-125 10126-127 10141-127 10127-125 10121-125 10137-126 10122-126 40102 40103 10138-600 10123-600 10138-124
RK10108EA 738.26 USD
RK10108 RK10121 RK10143 RK10105 RK10106 RK10107 RK10109 RK10111 RK10112 RK10120 RK10123 RK10124 RK10135 RK10138 RK10139 RK101411 RK10122 RK10125 RK10126 RK10127 RK10129 RK10136 RK10140 RK10142 RK10144 RK10128 RK10137 10058-074 10058-076 10058-078 10058-080 10058-090 10058-092 10058-094 10058-096 10058-152 10058-154 10058-158 10058-162 10058-308 10059-012 10803-454 10849-386 10849-564 10849-870 10850-172 10850-260 10850-264 10850-266 10850-268 10850-270 10850-374 10850-570 10850-578 10850-594 10850-606 10850-610 10850-628 10850-636 10850-638 10850-650 10850-730 10850-748 10850-754 10850-758 10850-760 10850-822 10850-824 10852-072 10852-092 75850-486 75851-578 75851-582 75851-618
Rtx®-1 GC Columns, Restek
Chromatography Columns
These general-purpose columns are designed for solvent impurities, PCB congeners, simulated distillation, drugs of abuse, gases, natural gas odorants, sulfur compounds, essential oils, hydrocarbons, semi-volatiles, pesticides, and oxygenates.
  • Low bleed levels
  • Longer column lifetime
  • Short conditioning times
Columns exhibit long lifetimes and very low bleed at high operating temperatures. A proprietary synthesis process eliminates residual catalysts that could cause degradation and increase bleed. Phase composition: crossbonded 100%More Product Information