VWR® Loading Dye Base 50X
Supplier: VWR
1B1812-25ML
10836-726EA 71.84 USD
10836-726
VWR® Loading Dye Base 50X
Stains Electrophoresis Stains
Loading Dye Base, 50X is used for custom loading buffer preparation.
  • Custom loading buffer preparation
  • Concentrated solution containing patent blue
  • Eliminates the need to weigh messy dye powders
Loading Dye Base, 50X is used for custom loading buffer preparation. The Loading Dye Base is a concentrated solution containing patent blue, bromophenol blue and amaranth tracking dyes, which migrate at approximately 4,000 bp, 400 bp and 10 bp, on a 1% agarose gel, respectively. Using Loading Dye Base eliminates the need to weigh messy dye powders.