MyCapLC™ Kits, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
2251211 2251310 2251311 2251151 2251251 2251351 2251130 2251131 2251230 2251110 2251111 2251210 2251231 2251330 2251331
10830-308EA 728.26 USD
10830-308 10830-310 10830-312 10830-314 10830-316 10830-318 10830-320 10830-322 10830-324 10830-706 10830-708 10830-710 10830-712 10830-714 10830-716
MyCapLC™ Kits, Trajan Scientific and Medical
Chromatography Columns
Customize CapLC for your application using the empty hardware provided.
  • DIY capillary column packing, no special intrumentation required
  • Make the columns that you want
  • Perfect for your life sciences analysis
  • Use the C18 media provided or use your own media
MyCapLC Kit enables you to create your own CapLC for higher resolution analysis compared to traditional HPLC. Includes simple packing instructions and you can easily pack your own capillary columns with speed in a matter of minutes using your own HPLC pump.
Capillary Liquid Chromatography (CapLC)More Product Information