8812 Series ESD Jacket, Transforming Technologies
Supplier: Transforming Technologies
JKC8809BG JKC8804BG JKC8803BG JKC8805BG JKC8806LB JKC8807LB JKC8808LB JKC8809LB JKC8804LB JKC8803LB JKC8802LB JKC8805LB JKC8806WH JKC8807WH JKC8808WH JKC8809WH JKC8804WH JKC8803WH JKC8802WH JKC8806SPBG JKC8807SPBG JKC8808SPBG JKC8809SPBG JKC8804SPBG JKC8803SPBG JKC8802SPBG JKC8805SPBG JKC8806SPLB JKC8807SPLB JKC8808SPLB JKC8809SPLB JKC8804SPLB JKC8803SPLB JKC8802SPLB JKC8805SPLB JKC8806SPWH JKC8807SPWH JKC8808SPWH JKC8809SPWH JKC8804SPWH JKC8803SPWH JKC8802SPWH JKC8805SPWH JKV8826BG JKV8827BG JKV8828BG JKV8829BG JKV8824BG JKV8823BG JKV8822BG JKV8825BG JKV8826LB JKV8827LB JKV8828LB JKV8829LB JKV8824LB JKV8823LB JKV8822LB JKV8825LB JKV8826WH JKV8827WH JKV8828WH JKV8829WH JKV8824WH JKV8823WH JKV8822WH JKV8825WH JKV8826SPBG JKV8827SPBG JKV8828SPBG JKV8829SPBG JKV8824SPBG JKV8823SPBG JKV8822SPBG JKV8825SPBG JKV8826SPLB JKV8827SPLB JKV8828SPLB JKV8829SPLB JKV8824SPLB JKV8823SPLB JKV8822SPLB JKV8825SPLB JKV8826SPWH JKV8827SPWH JKV8828SPWH JKV8829SPWH JKV8824SPWH JKV8823SPWH JKV8822SPWH JKV8825SPWH JKC8805WH JKC8802BG JKC8806BG JKC8807BG JKC8808BG
10828-588EA 96.91 USD
10828-588 10828-590 10828-592 10828-594 10828-634 10828-636 10828-638 10828-640 10828-642 10828-658 10828-670 10828-672 10828-734 10828-736 10828-738 10828-740 10828-742 10828-744 10828-746 10828-846 10828-848 10828-850 10828-852 10828-854 10828-856 10828-858 10828-860 10828-878 10828-880 10828-882 10828-884 10828-886 10828-888 10828-890 10828-892 10828-918 10828-920 10828-922 10828-924 10828-926 10828-928 10828-930 10828-932 10828-960 10828-962 10828-964 10828-966 10828-968 10828-970 10828-972 10828-974 10828-976 10828-978 10828-980 10828-982 10828-984 10828-986 10828-988 10828-990 10828-992 10828-994 10828-996 10828-998 10829-000 10829-002 10829-004 10829-006 10829-008 10829-010 10829-012 10829-014 10829-016 10829-018 10829-020 10829-022 10829-024 10829-026 10829-028 10829-030 10829-032 10829-034 10829-036 10829-038 10829-040 10829-042 10829-044 10829-046 10829-048 10829-050 10829-052 10829-054 10829-070 10829-140 10829-166 10829-168 10829-170
8812 Series ESD Jacket, Transforming Technologies
Workwear
The 8812 Series jackets from Transforming Technologies feature a patented hip-to-cuff feature for hands-free grounding
These garments are fabricated from lightweight 88% polyester and 12% carbon fabric for unmatched operator comfort. Superior workmanship provides reliable panel to panel continuity. The 8812 Series jackets maintains consistent continuity readings for up to 100 wash cycles.
Order Now
SPECIFICATIONS
Environmentally Preferable
Garment application Laboratory
Risk area ESD
Gender Unisex
Sleeve or leg length Long
Fabric 88% polyester and 12% carbon fabric