VWR® Plus Series Freestanding Countertop and Undercounter Freezers
Supplier: VWR International
10819-662 10819-664 10819-892 10834-034 76210-390 76210-392
10819-662EA 1170.44 USD
10819-662 10819-664 10819-892 10834-034 76210-390 76210-392
VWR® Plus Series Freestanding Countertop and Undercounter Freezers
Freezers Laboratory / General Purpose Freezers
These undercounter and countertop freezers provide lab quality cold storage in a space-saving design.
  • Integrated temperature alarms
  • Keyed door locks
  • Forced draft circulation
  • Precise microprocessor temperature control
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
Freestanding units require a 4-inch margin on all sides for adequate ventilation. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Select models offer auto defrost cycles for convenient defrosting. For lower temperatures, -30 and -40More Product Information