Chondroitin sulfate A sodium salt 90%

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium chondroitin sulphate, Chondroitin-4-sulphate, Chondroitin 4-sulphate sodium

J66156-06 J66156-14 J66156-22
10814-454EA 37.73 USD
10814-454 10814-456 10814-458
Chondroitin sulfate A sodium salt 90%
Chondroitin sulfate A sodium salt

Purity: 90% CAS No.: 39455-18-0 Molecular Formula: (C14H19NO14SNa2)n Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00130792

Formula: (C₁₄H₁₉NO₁₄SNa₂)n
Storage Temperature: Refrigerator
MDL Number: MFCD00130792
CAS Number: 39455-18-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR