UltraSOLV® 1700 Series Swabs, Pad Tip, Foamtec
Supplier: Foamtec International
HT1701-500
10811-142CS 4806.02 USD
10811-142
UltraSOLV® 1700 Series Swabs, Pad Tip, Foamtec
Swabs Swabs for Cleaning
This high-absorbancy, low-extractable foam head swab is ideal for semiconductor and electric assembly applications
Swab cleans, absorbs and removes contaminants from the surfaces of critical electronic devices. Polypropylene handle.
Información para pedidos: Swabs are packaged and sealed in five polypropylene mesh bags of 100 swabs each, and placed in a clear, polyethylene, cleanroom bag.