Anti-EGI Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-AZ7
10802-594EA 635.22 USD
10802-594
Anti-EGI Rabbit Polyclonal Antibody
Antibodies
EIG121 (Estrogen-induced gene 121 protein) is thought to play a role as a marker of hyperestrogenic state and estrogen-related type I endometrial carcinoma. It belongs to the UPF0577 family. It is a 1013 amino acid single-pass transmembrane protein that, though expressed in normal endometrium, is overexpressed in endometriod tumors. As an evolutionarily conserved gene, EIG121 is also expressed during early xenopus development, showing maximum expression at the gastrula stage.

Recommended Dilutions: ELISA: 1:10,000; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1 ?g/mL; contains 0.More Product Information
Order Now

SPECIFICATIONS

Antigen Symbol EGI
Environmentally Preferable
Antigen Name epilepsy, generalized, idiopathic
Antigen Synonyms EIG|EIG1
Antibody Type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 1957
Isotype
Western Blot Yes
ELISA Yes
ImmunoFluorescence Yes
Epitope C-Terminal
Immunogen Anti-EIG121 antibody was prepared from whole rabbit serum produced by repeated immunizations with an 18 amino acid synthetic peptide near the C-terminus of human EIG121.
Format IgG
Form Liquid (sterile filtered)
Purification Anti-EIG121 Antibody was affinity purified from monospecific antiserum by immunoaffinity chromatography. At least four isoforms of EIG121 known to exist. EIG121 antibody is predicted to not cross-react with other UPF0577 protein family members.
Concentration 1 mg/mL by UV absorbance at 280 nm
Shipping Temperature Dry Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR