Cryo.s™ Cryogenic Storage Vials, Polypropylene, Sterile, Greiner Bio-One
Supplier: Greiner Bio-One
121261 121263 121277 121278 121279 121280 122261 122263 122278 122279 123261 123263 123277 123278 123279 123280 124261 124263 124273 124274 124275 124276 126261 126263 126277 126278 126280 127261 127277 127279 127280 122277 122280 126279 127263 127278 122272
82050-154CS 269.81 USD
82050-154 82050-156 82050-158 82050-160 82050-162 82050-164 82050-166 82050-168 82050-172 82050-174 82050-178 82050-180 82050-182 82050-184 82050-186 82050-188 82050-190 82050-192 82050-194 82050-196 82050-198 82050-200 82050-204 82050-206 82050-208 82050-210 82050-214 82050-216 82050-220 82050-224 82050-226 82050-170 82050-176 82050-212 82050-218 82050-222 10799-204
Cryo.s™ Cryogenic Storage Vials, Polypropylene, Sterile, Greiner Bio-One
Vials
Designed for short- and long-term storage of samples and cells, these cryogenic storage vials are made from virgin polypropylene resin for purity, clarity, and consistency.
  • Nonpyrogenic and RNase- and DNase-free to <0.06 EU/ml
Cryo.s™ vials are available with internal or external thread. The internal thread format features a silicone gasket and is ideally suited for storage in the vapor phase of liquid nitrogen, at temperatures from –196 to 121 °C.
Información de suministro: All vials are supplied with caps. Each case includes cap inserts: 150More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable