Cimarec+™ Magnetic Stirrers, Thermo Scientific
Supplier: Thermo Fisher Scientific
S88857100 S88857104 S88850100 S88854100
10790-818EA 474.66 USD
10790-818 10790-824 10790-832 10791-582
Cimarec+™ Magnetic Stirrers, Thermo Scientific
Stirrers Magnetic Stirrers
The Thermo Scientific Cimarec+™ series of digital magnetic stirrers are designed to provide precise stirring control for routine protocols
All units offer digital displays of rpm. Patented StirTrac™ technology provides innovative engineering for slow-speed stirring, consistent speed control, and strong magnetic coupling. The Cimarec+™ series features a raised display designed to protect electronics from spills. Units include one magnetic stirring bar. Optional splash guard protection shields are available separately.
Corresponding magnetic hotplate stirrers (stirring and heating 10790-8More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable