OptiTDS™, Adrenal Tissue Dissociation System, CHI Scientific
Supplier: CHI Scientific
4-20208 4-20209 4-20201 4-20202 4-20203 4-20204 4-20205 4-20206 4-20207
10783-150EA 420.64 USD
10783-150 10783-152 10786-080 10786-220 10786-222 10786-224 10786-226 10786-228 10786-230
OptiTDS™, Adrenal Tissue Dissociation System, CHI Scientific
Protein Extraction Protein Extraction Kits
OptiTDS™ is a proprietary Tissue Dissociation system which enzymatically digests extracellular connective fibers without harming the cells themselves
Stability/Storage: The reagents contained in this system are stable at room temperatures for normal shipping periods, but the kit should be used immediately or the package should be stored at -20oC upon arrival. The reagents contained in this kit are stable and effective for up to 6 months at -20oC.
Each OptiTDS™ system contains concentrated tissue dissociation enzymes and reagents which are added to an included buffer system to produce a working solution that is capable of treating 3-5 g of tissue. Each tissue dissociation system is customized to be tissue-specific and cell-specific to obtain the best results for each type of primary cell.