Human Recombinant BMP-4 (from HeLa cells)
Supplier: Peprotech
120-05-1UG 120-05-5UG 120-05-100UG 120-05-500UG 120-05-1MG
10779-134EA 143.88 USD
10779-134 10779-136 10779-138 10779-140 10779-142
Human Recombinant BMP-4 (from HeLa cells)
Proteins and Peptides
Bone morphogenetic proteins (BMPs) constitute a subfamily within the TGF-β superfamily of structurally related signaling proteins. Members of this superfamily are widely distributed throughout the body, and are involved in diverse physiological processes during both pre- and postnatal life. Like BMP-7, BMP-4 is involved in the development and maintenance of bone and cartilage. Reduced expression of BMP-4 is associated with a number of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Recombinant Human BMP-4, expressed in
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.