Human Recombinant IL36G (from E. coli)
Supplier: Peprotech
AF-200-36G-2UG AF-200-36G-10UG AF-200-36G-50UG AF-200-36G-100UG AF-200-36G-250UG AF-200-36G-500UG AF-200-36G-1MG
10778-430EA 183.06 USD
10778-430 10778-432 10778-434 10778-436 10778-438 10778-440 10778-442
Human Recombinant IL36G (from E. coli)
Proteins and Peptides
The IL-1 family is comprised of 11 structurally related ligands, including recently re-named IL-36 α (IL-1F6), β (IL-1F8) and γ (IL-1F9). IL-36γ is highly expressed in psoriatic plaques and in tissues containing epithelial cells. IL-36γ signals through the IL-1Rrp2 (IL-1R6) receptor, which is primarily expressed on certain dendritic cells. The interaction of the IL-1Rrp2 receptor with IL-36 ligands induces dendritic cell maturation and activation. IL-36γ also functions as an agonist of NF-κB, and can stimulate the inflammatory response in bronchial epithelial cells. Recombinant Human IL-36γ is a 17.0 kDa protein containing 152 amino acid residues. Manufactured using all Animal-Free reagents.
  • Carrier free
  • Animal-free
SMCKPITGTI NDLNQQVWTL QGQNLVAVPR SDSVTPVTVA VITCKYPEAL EQGRGDPIYL GIQNPEMCLY CEKVGEQPTL QLKEQKIMDL YGQPEPVKPF LFYRAKTGRT STLESVAFPD WFIASSKRDQ PIILTSELGK SYNTAFELNI ND
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.