Human Recombinant GDF-3 (from E. coli)
Supplier: Peprotech
AF-120-22-5UG AF-120-22-20UG AF-120-22-100UG AF-120-22-250UG AF-120-22-500UG AF-120-22-1MG
10778-186EA 183.06 USD
10778-186 10778-188 10778-190 10778-192 10778-194 10778-196
Human Recombinant GDF-3 (from E. coli)
Proteins and Peptides
GDF-3 is a member of the TGF-β superfamily of growth and differentiation factors, and is highly homologous to GDF-9. Unlike most TGF-β family members, GDF-3 and GDF-9 are not disulfide-linked dimers. GDF-3 is expressed in adult bone marrow, spleen, thymus, and adipose tissue. The expression of GDF-3 is upregulated in high-fat-fed wild-type FABP4/aP2 null mice and was associated with obesity, but not with the related hyperglycemia/hyperinsulinemia that characterizes Type 2 diabetes. Recombinant Human GDF-3 is a 26.0 kDa non-disulfide-linked homodimer containing two 114 amino acid polypeptide chains.
  • Animal-free
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.