Anti-MIP-1beta (CCL4) Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P38BBT-25UG 500-P38BBT-50UG
10771-294EA 354.54 USD
10771-294 10771-296 10771-298
Anti-MIP-1beta (CCL4) Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human MIP-1β (CCL4). Anti­Human MIP-1β (CCL4)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: MIP-1beta (CCL4)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen Symbol CCL4
Antigen Name MIP-1beta
Environmentally Preferable
Antibody Type Secondary
Clonality Polyclonal
Conjugation Biotin
Reactivity Human
Host Rabbit
Isotype IgG
Western Blot Yes
ELISA Yes
Immunogen Recombinant Human MIP-1β (CCL4)
Form Lyophilized
Molecular Weight 7.6 kDa
Purification Recombinant Human MIP-1β (CCL4)
Storage Buffer See Product Data Sheet for more information.
Storage Temperature Store at –20 °C prior to reconstitution.
Shipping Temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR