LaboPlast® Bio / SteriPlast® Bio, Sampling scoops, Bürkle
Supplier: BURKLE INC
5379-1007 5379-0007 5379-1005 5379-0005 5379-1003 5379-0003 5379-1001 5379-0001 5379-0024 5379-1024 5379-0025 5379-1025
10769-668PK 56.18 USD
10769-668 10769-718 10769-912 10769-914 10770-092 10770-094 10770-202 10770-204 76501-364 76501-366 76501-374 76501-376
LaboPlast® Bio / SteriPlast® Bio, Sampling scoops, Bürkle
Scoops
These disposable scoops are manufactured from bio-plastic "Green PE" which is entirely made of renewable resources
  • "Green PE" is 100% recyclable, made without fossil raw materials
  • Available non sterile (LaboPlast Bio) or gamma radiation sterilised (SteriPlast Bio)
  • Produced in a cleanroom environment
  • Individually packed
They are ideal for the sampling of powder, granules and pastes.
Certificaciones: Complies with EU food requirements and FDA regulations.