Anti-Granzyme B Rat Monoclonal Antibody [clone: 12F9B65]

Supplier: Biolegend
662801 662802
10763-628EA 113.7 USD
10763-628 10763-630
Anti-Granzyme B Rat Monoclonal Antibody [clone: 12F9B65]
Antibodies
Purified anti-Granzyme B [12F9B65]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: WB; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.1 - 0.5 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Granzyme B
Clonality: Monoclonal
Clone: 12F9B65
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

SPECIFICATIONS

Antigen Name Granzyme B
Environmentally Preferable
Antigen Synonyms CCP1|GZMB|cytotoxic T-lymphocyte-associated serine esterase 1|Asp-ase|Granzyme 2|Aspase
Antibody Type Primary
Clonality Monoclonal
Clone 12F9B65
Reactivity Mouse
Host Rat
Gene ID 14939
Isotype IgG2a kappa
Western Blot Yes
Immunogen Partial mouse Granzyme B recombinant protein (17-247 aa) expressed in E. coli.
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Purification The antibody was purified by affinity chromatography.
Concentration 0.50 mg/mL
Storage Temperature The antibody solution should be stored undiluted between 2 °C and 8 °C.
Shipping Temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR