Anti-Tubulin Beta 3 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: AA10]

Supplier: Biolegend
657405 657406
10763-570EA 198.1 USD
10763-570 10763-572
Anti-Tubulin Beta 3 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: AA10]
Antibodies
Alexa Fluor® 647 anti-Tubulin Beta 3 (TUBB3) [AA10]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse, Rat; Apps: ICFC, IHC-F; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.06 μg per million cells in 100 μl volume. For immunohistochemical staining on frozen tissue sections, the suggested use is 1.25-5 μg/mL. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen:More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen Name Tubulin Beta 3
Antigen Synonyms β-3 Tubulin|Tubulin beta3|Tubulin Beta III|Neuronal class III beta-tubulin|TUBB3|β3-tub
Antibody Type Primary
Clonality Monoclonal
Conjugation Alexa Fluor® 647
Clone AA10
Reactivity Human,Rat,Mouse
Host Mouse
Gene ID 10381
Isotype IgG2a kappa
ImmunoChemistry Yes
Immunogen Fusion protein
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Purification The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 647 under optimal conditions.
Concentration 0.50 mg/mL
Storage Temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping Temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR